missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl (1S)-1-Phenyl-3,4-dihydroisoquinoline-2(1H)-carboxylate, Mikromol
Click to view available options
Quantity:
25 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
Ethyl (1S)-1-Phenyl-3,4-dihydroisoquinoline-2(1H)-carboxylate
Specifications
Specifications
| Chemical Name or Material | Ethyl (1S)-1-Phenyl-3,4-dihydroisoquinoline-2(1H)-carboxylate |
| CAS | 180468-42-2 |
| Analyte or Component Names | Ethyl (1S)-1-Phenyl-3,4-dihydroisoquinoline-2(1H)-carboxylate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H19 N O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | (1S)-3,4-Dihydro-1-phenyl-2(1H)-isoquinolinecarboxylic acid ethyl ester, 2(1H)-Isoquinolinecarboxylic acid, 3,4-dihydro-1-phenyl-, ethyl ester, (S)-, (S)-1-Phenyl-3,4-dihydro-1H-isoquinolinecarboxylic acid ethyl ester, Ethyl (1S)-1-phenyl-1,2,3,4-tetrahydroisoquinoline-2-carboxylate, Ethyl (1S)-3,4-dihydro-1-phenyl-2(1H)-isoquinolinecarboxylate, Ethyl (S)-1-phenyl-1,2,3,4-tetrahydro-2-isoquinolinecarboxylate, Ethyl (S)-1-phenyl-1,2,3,4-tetrahydroisoquinoline-2-carboxylate |
| InChI Formula | InChI=1S/C18H19NO2/c1-2-21-18(20)19-13-12-14-8-6-7-11-16(14)17(19)15-9-4-3-5-10-15/h3-11,17H,2,12-13H2,1H3/t17-/m0/s1 |
| SMILES | CCOC(=O)N1CCc2ccccc2[C@@H]1c3ccccc3 |
| IUPAC Name | ethyl (1S)-1-phenyl-3,4-dihydro-1H-isoquinoline-2-carboxylate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction