missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Febuxostat Ethyl Ester (2-[3-Cyano-4-(2-methylpropoxy)phenyl]-4-methyl-5-thiazolecarboxylic Acid Ethyl Ester), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Febuxostat Ethyl Ester (2-[3-Cyano-4-(2-methylpropoxy)phenyl]-4-methyl-5-thiazolecarboxylic Acid Ethyl Ester)
Specifications
Specifications
| Chemical Name or Material | 2-[3-Cyano-4-(2-methylpropoxy)phenyl]-4-methyl-5-thiazolecarboxylic Acid Ethyl Ester |
| CAS | 160844-75-7 |
| Analyte or Component Names | 2-[3-Cyano-4-(2-methylpropoxy)phenyl]-4-methyl-5-thiazolecarboxylic Acid Ethyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H20 N2 O3 S |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-[3-Cyano-4-(2-methylpropoxy)phenyl]-4-methyl-5-thiazolecarboxylic Acid Ethyl Ester, Ethyl 2-(3-cyano-4-isobutoxyphenyl)-4-methyl-5-thiazolecarboxylate |
| InChI Formula | InChI=1S/C18H20N2O3S/c1-5-22-18(21)16-12(4)20-17(24-16)13-6-7-15(14(8-13)9-19)23-10-11(2)3/h6-8,11H,5,10H2,1-4H3 |
| SMILES | CCOC(=O)c1sc(nc1C)c2ccc(OCC(C)C)c(c2)C#N |
| IUPAC Name | ethyl 2-[3-cyano-4-(2-methylpropoxy)phenyl]-4-methyl-1,3-thiazole-5-carboxylate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction