missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fenoprofen 1,2,3-Propanetriol Esters (Mixture of Regio- and Stereoisomers), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Fenoprofen 1,2,3-Propanetriol Esters (Mixture of Regio- and Stereoisomers)
Specifications
Specifications
| Chemical Name or Material | Fenoprofen 1,2,3-Propanetriol Esters (Mixture of Regio- and Stereoisomers) |
| Analyte or Component Names | Fenoprofen 1,2,3-Propanetriol Esters (Mixture of Regio- and Stereoisomers) |
| Shipping Condition | Room Temperature |
| Molecular Formula | 2 C18 H20 O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Fenoprofen 1,2,3-Propanetriol Esters (Mixture of Regio- and Stereoisomers), Fenoprofen 1,2,3-Propanetriol Ester (Mixture of Isomers) |
| InChI Formula | InChI=1S/2C18H20O5/c1-13(18(21)22-12-15(20)11-19)14-6-5-9-17(10-14)23-16-7-3-2-4-8-16;1-13(18(21)23-17(11-19)12-20)14-6-5-9-16(10-14)22-15-7-3-2-4-8-15/h2-10,13,15,19-20H,11-12H2,1H3;2-10,13,17,19-20H,11-12H2,1H3 |
| SMILES | CC(C(=O)OCC(O)CO)c1cccc(Oc2ccccc2)c1.CC(C(=O)OC(CO)CO)c3cccc(Oc4ccccc4)c3 |
| IUPAC Name | 1,3-dihydroxypropan-2-yl 2-(3-phenoxyphenyl)propanoate;2,3-dihydroxypropyl 2-(3-phenoxyphenyl)propanoate |
| Molecular Weight (g/mol) | 632.7 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction