missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fenoprofen 2,3-Butylene Glycol Ester, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Fenoprofen 2,3-Butylene Glycol Ester
Specifications
Specifications
| Chemical Name or Material | Fenoprofen 2,3-Butylene Glycol Ester |
| Analyte or Component Names | Fenoprofen 2,3-Butylene Glycol Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H22 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Fenoprofen 2,3-Butylene Glycol Ester |
| InChI Formula | InChI=1S/C19H22O4/c1-13(19(21)22-15(3)14(2)20)16-8-7-11-18(12-16)23-17-9-5-4-6-10-17/h4-15,20H,1-3H3 |
| SMILES | CC(O)C(C)OC(=O)C(C)c1cccc(Oc2ccccc2)c1 |
| IUPAC Name | 3-hydroxybutan-2-yl 2-(3-phenoxyphenyl)propanoate |
| Molecular Weight (g/mol) | 314.38 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction