missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ganciclovir Mono-N-methyl Valinate Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Ganciclovir Mono-N-methyl Valinate Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Ganciclovir Mono-N-methyl Valinate Hydrochloride |
| Analyte or Component Names | Ganciclovir Mono-N-methyl Valinate Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H24 N6 O5 . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | (2-(RS)-[(guanin-9-yl)methoxy]-3-hydroxypropyl N-methyl-L-valinate hydrochloride) |
| InChI Formula | InChI=1S/C15H24N6O5.ClH/c1-8(2)10(17-3)14(24)25-5-9(4-22)26-7-21-6-18-11-12(21)19-15(16)20-13(11)23;/h6,8-10,17,22H,4-5,7H2,1-3H3,(H3,16,19,20,23);1H/t9?,10-;/m0./s1 |
| SMILES | Cl.CN[C@@H](C(C)C)C(=O)OCC(CO)OCn1cnc2C(=O)NC(=Nc12)N |
| IUPAC Name | [2-[(2-amino-6-oxo-1H-purin-9-yl)methoxy]-3-hydroxypropyl] (2S)-3-methyl-2-(methylamino)butanoate;hydrochloride |
| Molecular Weight (g/mol) | 404.85 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction