missing translation for 'onlineSavingsMsg'
Learn More

Glycodeoxycholic Acid, Mikromol™

Product Code. 17900676
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 17900676

Brand: Mikromol MM3875.020025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Glycodeoxycholic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glycodeoxycholic Acid
CAS 360-65-6
Analyte or Component Names Glycodeoxycholic Acid
Shipping Condition Room Temperature
Molecular Formula C26 H43 N O5
Certifications/Compliance ISO 9001
Synonym Glycine, N-[(3α,5β,12α)-3,12-dihydroxy-24-oxocholan-24-yl]-, Glycine, N-(3α,12α-dihydroxy-5β-cholan-24-oyl)- (8CI), Glycodeoxycholic acid (6CI,7CI), 5β-Cholan-24-amide, N-(carboxymethyl)-3α,12α-dihydroxy- (8CI), Cholane, glycine deriv., N-[(3α,5β,12α)-3,12-Dihydroxy-24-oxocholan-24-yl]glycine, 3α,12α-Dihydroxy-5β-cholanic acid-24-glycine, Deoxycholic acid glycine conjugate, Deoxycholylglycine, Deoxyglycocholic acid, Glycodesoxycholic acid, Glycyldeoxycholic acid
InChI Formula InChI=1S/C26H43NO5/c1-15(4-9-23(30)27-14-24(31)32)19-7-8-20-18-6-5-16-12-17(28)10-11-25(16,2)21(18)13-22(29)26(19,20)3/h15-22,28-29H,4-14H2,1-3H3,(H,27,30)(H,31,32)/t15-,16-,17-,18+,19-,20+,21+,22+,25+,26-/m1/s1
SMILES C[C@H](CCC(=O)NCC(=O)O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C
IUPAC Name 2-[[(4R)-4-[(3R,5R,8R,9S,10S,12S,13R,14S,17R)-3,12-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]acetic acid
Molecular Weight (g/mol) 449.62
Quantity 25 mg
Formula Weight 449.3141 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.