missing translation for 'onlineSavingsMsg'
Learn More
Learn More
iso-Miconazole (1-[1-(2,4-Dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
iso-Miconazole (1-[1-(2,4-Dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole)
Specifications
Specifications
| Chemical Name or Material | iso-Miconazole |
| CAS | 178813-04-2 |
| Analyte or Component Names | iso-Miconazole |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H14 Cl4 N2 O |
| Certifications/Compliance | ISO 9001 |
| Synonym | 1-[1-(2,4-Dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]-1H-imidazole, iso-Miconazole |
| InChI Formula | InChI=1S/C18H14Cl4N2O/c19-13-2-1-12(16(21)7-13)9-25-10-18(24-6-5-23-11-24)15-4-3-14(20)8-17(15)22/h1-8,11,18H,9-10H2 |
| SMILES | Clc1ccc(COCC(c2ccc(Cl)cc2Cl)n3ccnc3)c(Cl)c1 |
| IUPAC Name | 1-[1-(2,4-dichlorophenyl)-2-[(2,4-dichlorophenyl)methoxy]ethyl]imidazole |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction