missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Isopropyl Thiophene-2-carboxylate, Mikromol™
Description
Isopropyl Thiophene-2-carboxylate
Specifications
Specifications
| Chemical Name or Material | Isopropyl Thiophene-2-carboxylate |
| CAS | 111160-47-5 |
| Analyte or Component Names | Isopropyl Thiophene-2-carboxylate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H10 O2 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Isopropyl Thiophene-2-carboxylate, 2-Thiophenecarboxylic acid 1-methylethyl ester, Isopropyl 2-thienylcarboxylate |
| InChI Formula | InChI=1S/C8H10O2S/c1-6(2)10-8(9)7-4-3-5-11-7/h3-6H,1-2H3 |
| SMILES | CC(C)OC(=O)c1cccs1 |
| IUPAC Name | propan-2-yl thiophene-2-carboxylate |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?