missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mefenamic Acid Ethyl Ester, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Mefenamic Acid Ethyl Ester
Specifications
Specifications
| Chemical Name or Material | Mefenamic Acid Ethyl Ester |
| CAS | 1226-33-1 |
| Analyte or Component Names | Mefenamic Acid Ethyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H19 N O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Mefenamic Acid Ethyl Ester, 2-[(2,3-Dimethylphenyl)amino]-benzoic acid ethyl ester, Anthranilic acid, N-2,3-xylyl-, ethyl ester (7CI,8CI), Ethyl 2-[(2,3-dimethylphenyl)amino]benzoate, Ethyl mefenamate |
| InChI Formula | InChI=1S/C17H19NO2/c1-4-20-17(19)14-9-5-6-10-16(14)18-15-11-7-8-12(2)13(15)3/h5-11,18H,4H2,1-3H3 |
| SMILES | CCOC(=O)c1ccccc1Nc2cccc(C)c2C |
| IUPAC Name | ethyl 2-(2,3-dimethylanilino)benzoate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction