missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mequitazine Sulfone N-Oxide (Mequitazine N,S,S-Trioxide), Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Mequitazine Sulfone N-Oxide (Mequitazine N,S,S-Trioxide)
Specifications
Specifications
| Chemical Name or Material | Mequitazine Sulfone N-Oxide |
| Analyte or Component Names | Mequitazine Sulfone N-Oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H22 N2 O3 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Mequitazine Sulfone N-Oxide |
| InChI Formula | InChI=1S/C20H22N2O3S/c23-22-11-9-15(10-12-22)16(14-22)13-21-17-5-1-3-7-19(17)26(24,25)20-8-4-2-6-18(20)21/h1-8,15-16H,9-14H2 |
| SMILES | [O-][N+]12CCC(CC1)C(CN3c4ccccc4S(=O)(=O)c5ccccc35)C2 |
| IUPAC Name | 10-[(1-oxido-1-azoniabicyclo[2.2.2]octan-3-yl)methyl]phenothiazine 5,5-dioxide |
| Molecular Weight (g/mol) | 370.47 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction