missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-[2-[(2,6-Dichlorophenyl)amino]phenyl]acetate (Methyl Ester of Diclofenac), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Methyl 2-[2-[(2,6-Dichlorophenyl)amino]phenyl]acetate (Methyl Ester of Diclofenac)
Specifications
Specifications
| Chemical Name or Material | Diclofenac methyl ester |
| CAS | 15307-78-5 |
| Analyte or Component Names | Diclofenac methyl ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H13 Cl2 N O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Methyl [2-[(2,6-Dichlorophenyl)amino]phenyl]acetate, Methyl 2-[2-[(2,6-Dichlorophenyl)amino]phenyl]acetate, Aceclofenac Imp. B (EP), Methyl Ester of Diclofenac, Aceclofenac Impurity B |
| InChI Formula | InChI=1S/C15H13Cl2NO2/c1-20-14(19)9-10-5-2-3-8-13(10)18-15-11(16)6-4-7-12(15)17/h2-8,18H,9H2,1H3 |
| SMILES | COC(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl |
| IUPAC Name | methyl 2-[2-(2,6-dichloroanilino)phenyl]acetate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction