missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinecarboxylate, Mikromol
Click to view available options
Quantity:
25 mg
4 x 25 mg
Unit Size:
25mg
Description
Methyl 2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinecarboxylate
Specifications
Specifications
| Chemical Name or Material | Methyl 2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinecarboxylate |
| CAS | 121659-86-7 |
| Analyte or Component Names | Methyl 2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinecarboxylate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H16 F N O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 2-Cyclopropyl-4-(4-fluorophenyl)-3-quinolinecarboxylic Acid Methyl Ester, 2-Cyclopropyl-4-(4-fluorophenyl)quinoline-3-carboxylic acid methyl ester, 4-(4-Fluorophenyl)-2-cyclopropylquinoline-3-carboxylic acid methyl ester, Methyl 2-cyclopropyl-4-(4-fluorophenyl)-3-quinolinecarboxylate |
| InChI Formula | InChI=1S/C20H16FNO2/c1-24-20(23)18-17(12-8-10-14(21)11-9-12)15-4-2-3-5-16(15)22-19(18)13-6-7-13/h2-5,8-11,13H,6-7H2,1H3 |
| SMILES | COC(=O)c1c(nc2ccccc2c1c3ccc(F)cc3)C4CC4 |
| IUPAC Name | methyl 2-cyclopropyl-4-(4-fluorophenyl)quinoline-3-carboxylate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction