missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Methyl 3alpha,7beta-Dihydroxy-5beta-cholan-24-oate, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Methyl 3alpha,7beta-Dihydroxy-5beta-cholan-24-oate
Specifications
Specifications
| Chemical Name or Material | Ursodeoxycholic-1-methyl Ester |
| CAS | 10538-55-3 |
| Analyte or Component Names | Ursodeoxycholic-1-methyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C25 H42 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Chenodeoxycholic Acid Imp. G (EP), (3alpha,5beta,7beta)-3,7-Dihydroxycholan-24-oic acid methyl ester, 3alpha,7beta-Dihydroxy-5beta-cholan-24-oic acid methyl ester), 3alpha,7beta-Dihydroxy-5beta-cholanic acid methyl ester, Methyl 3alpha,7beta-dihydroxy-5beta-cholanoate, Methyl ursodeoxycholate, Ursodeoxycholic acid methyl ester, Chenodeoxycholic Acid Impurity G, Ursodeoxycholic Acid Impurity G |
| InChI Formula | InChI=1S/C25H42O4/c1-15(5-8-22(28)29-4)18-6-7-19-23-20(10-12-25(18,19)3)24(2)11-9-17(26)13-16(24)14-21(23)27/h15-21,23,26-27H,5-14H2,1-4H3/t15-,16+,17-,18-,19+,20+,21+,23+,24+,25-/m1/s1 |
| SMILES | COC(=O)CC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3[C@@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C |
| IUPAC Name | methyl (4R)-4-[(3R,5S,7S,8R,9S,10S,13R,14S,17R)-3,7-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction