missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Flubendazole EP Impurity A, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
Flubendazole EP Impurity A
Specifications
Specifications
| Chemical Name or Material | Methyl [5-[4-(Formylamino)benzoyl]-1H-benzimidazol-2-yl]carbamate |
| Analyte or Component Names | Methyl [5-[4-(Formylamino)benzoyl]-1H-benzimidazol-2-yl]carbamate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H14 N4 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Flubendazole Imp. A (EP), Flubendazole Impurity A |
| InChI Formula | InChI=1S/C17H14N4O4/c1-25-17(24)21-16-19-13-7-4-11(8-14(13)20-16)15(23)10-2-5-12(6-3-10)18-9-22/h2-9H,1H3,(H,18,22)(H2,19,20,21,24) |
| SMILES | COC(=O)Nc1nc2cc(ccc2[nH]1)C(=O)c3ccc(NC=O)cc3 |
| IUPAC Name | methyl N-[5-(4-formamidobenzoyl)-1H-benzimidazol-2-yl]carbamate |
| Molecular Weight (g/mol) | 338.32 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction