missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Montelukast Dicyclohexylamine, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Montelukast Dicyclohexylamine
Specifications
Specifications
| Chemical Name or Material | Montelukast Dicyclohexylamine |
| CAS | 577953-88-9 |
| Analyte or Component Names | Montelukast Dicyclohexylamine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C35 H36 Cl N O3 S . C12 H23 N |
| Certifications/Compliance | ISO 17025 |
| Synonym | Montelukast Dicyclohexylamine, Cyclopropaneacetic acid, 1-[[[(1R)-1-[3-[(1E)-2-(7-chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]thio]methyl]-, compd. with N-cyclohexylcyclohexanamine (1:1) (9CI), 2-[1-[[[(1R)-1-[3-[(1E)-2-(7-Chloro-2-quinolinyl)ethenyl]phenyl]-3-[2-(1-hydroxy-1-methylethyl)phenyl]propyl]thio]methyl]cyclopropyl]acetic acid dicyclohexylamine salt, Montelukast dicyclohexylamine salt |
| InChI Formula | InChI=1S/C35H36ClNO3S.C12H23N/c1-34(2,40)30-9-4-3-7-25(30)13-17-32(41-23-35(18-19-35)22-33(38)39)27-8-5-6-24(20-27)10-15-29-16-12-26-11-14-28(36)21-31(26)37-29;1-3-7-11(8-4-1)13-12-9-5-2-6-10-12/h3-12,14-16,20-21,32,40H,13,17-19,22-23H2,1-2H3,(H,38,39);11-13H,1-10H2/b15-10+;/t32-;/m1./s1 |
| SMILES | CC(C)(O)c1ccccc1CC[C@@H](SCC2(CC(=O)O)CC2)c3cccc(\C=C\c4ccc5ccc(Cl)cc5n4)c3.C6CCC(CC6)NC7CCCCC7 |
| IUPAC Name | 2-[1-[[(1R)-1-[3-[(E)-2-(7-chloroquinolin-2-yl)ethenyl]phenyl]-3-[2-(2-hydroxypropan-2-yl)phenyl]propyl]sulfanylmethyl]cyclopropyl]acetic acid;N-cyclohexylcyclohexanamine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction