missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[(1RS)-1-Methyl-2-phenylethyl]prop-2-yn-1-amine Hydrochloride (rac-Demethylselegiline Hydrochloride), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
N-[(1RS)-1-Methyl-2-phenylethyl]prop-2-yn-1-amine Hydrochloride (rac-Demethylselegiline Hydrochloride)
Specifications
Specifications
| Chemical Name or Material | rac-N-Desmethylselegiline Hydrochloride |
| CAS | 2588-96-7 |
| Analyte or Component Names | rac-N-Desmethylselegiline Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H15 N . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | N-[(1RS)-1-Methyl-2-phenylethyl]prop-2-yn-1-amine Hydrochloride, Selegiline Hydrochloride Imp. D (EP) as Racemate Hydrochloride, rac-Demethylselegiline Hydrochloride, rac-N-Desmethylselegiline Hydrochloride, Selegiline Hydrochloride Imp. D (EP), Selegiline Imp. D (EP), Selegiline Hydrochloride Impurity D as Racemate Hydrochloride, Selegiline Impurity D as Racemate Hydrochloride |
| InChI Formula | InChI=1S/C12H15N.ClH/c1-3-9-13-11(2)10-12-7-5-4-6-8-12;/h1,4-8,11,13H,9-10H2,2H3;1H |
| SMILES | Cl.CC(Cc1ccccc1)NCC#C |
| IUPAC Name | 1-phenyl-N-prop-2-ynylpropan-2-amine;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction