Learn More
Nimesulide EP Impurity A, Mikromol™
Product Code. 30050231
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. 30050231
Brand: Mikromol MM0378.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Nimesulide EP Impurity A
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material N-(2,4-Dinitro-6-phenoxyphenyl)methanesulphonamide
CAS 51765-56-1
Analyte or Component Names N-(2,4-Dinitro-6-phenoxyphenyl)methanesulphonamide
Shipping Condition Room Temperature
Molecular Formula C13 H11 N3 O7 S
Certifications/Compliance ISO 17025
Synonym N-(2,4-Dinitro-6-phenoxyphenyl)methanesulphonamide, Nimesulide Imp. A (EP), Nimesulide Impurity A
InChI Formula InChI=1S/C13H11N3O7S/c1-24(21,22)14-13-11(16(19)20)7-9(15(17)18)8-12(13)23-10-5-3-2-4-6-10/h2-8,14H,1H3
SMILES CS(=O)(=O)Nc1c(Oc2ccccc2)cc(cc1[N+](=O)[O-])[N+](=O)[O-]
IUPAC Name N-(2,4-dinitro-6-phenoxyphenyl)methanesulfonamide
Molecular Weight (g/mol) 353.31
Quantity 25 mg
Formula Weight 353.0318 g/mol