missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-[3-(morpholin-4-yl)propoxy]-N-[3-(morpholin-4-yl)propyl]quinazolin-4-amine, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-[3-(morpholin-4-yl)propoxy]-N-[3-(morpholin-4-yl)propyl]quinazolin-4-amine
Specifications
Specifications
| Chemical Name or Material | N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-[3-(morpholin-4-yl)propoxy]-N-[3-(morpholin-4-yl)propyl]quinazolin-4-amine |
| CAS | 1502829-45-9 |
| Analyte or Component Names | N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-[3-(morpholin-4-yl)propoxy]-N-[3-(morpholin-4-yl)propyl]quinazolin-4-amine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C29 H37 Cl F N5 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | 4-Quinazolinamine, N-(3-chloro-4-fluorophenyl)-7-methoxy-6-[3-(4-morpholinyl)propoxy]-N-[3-(4-morpholinyl)propyl]-, N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-[3-(4-morpholinyl)propoxy]-N-[3-(4-morpholinyl)propyl]-4-quinazolinamine, N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-[3-(morpholin-4-yl)propoxy]-N-[3-(morpholin-4-yl)propyl]quinazolin-4-amine |
| InChI Formula | InChI=1S/C29H37ClFN5O4/c1-37-27-20-26-23(19-28(27)40-13-3-7-35-11-16-39-17-12-35)29(33-21-32-26)36(22-4-5-25(31)24(30)18-22)8-2-6-34-9-14-38-15-10-34/h4-5,18-21H,2-3,6-17H2,1H3 |
| SMILES | COc1cc2ncnc(N(CCCN3CCOCC3)c4ccc(F)c(Cl)c4)c2cc1OCCCN5CCOCC5 |
| IUPAC Name | N-(3-chloro-4-fluorophenyl)-7-methoxy-6-(3-morpholin-4-ylpropoxy)-N-(3-morpholin-4-ylpropyl)quinazolin-4-amine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction