missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-[4-[[(1-Methylethyl)amino]acetyl]phenyl]methanesulphonamide Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
N-[4-[[(1-Methylethyl)amino]acetyl]phenyl]methanesulphonamide Hydrochloride
Specifications
Specifications
| Chemical Name or Material | N-[4-[((1-Methylethyl)amino)acetyl]phenyl]-methanesulphonamide Hydrochloride |
| CAS | 5576-49-8 |
| Analyte or Component Names | N-[4-[((1-Methylethyl)amino)acetyl]phenyl]-methanesulphonamide Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C12 H18 N2 O3 S . Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Sotalol Hydrochloride Imp. B (EP), Sotalol Imp. B (EP), Sotalol USP Related Compound A, Sotalol USP RC A, N-[4-[[(1-Methylethyl)amino]acetyl]phenyl]-methanesulphonamide Hydrochloride, Sotalol Hydrochloride Imp. B (EP) as Hydrochloride, Sotalol Related Compound A as Hydrochloride, Sotalol Hydrochloride Impurity B as Hydrochloride, Sotalol Impurity B as Hydrochloride |
| InChI Formula | InChI=1S/C12H18N2O3S.ClH/c1-9(2)13-8-12(15)10-4-6-11(7-5-10)14-18(3,16)17;/h4-7,9,13-14H,8H2,1-3H3;1H |
| SMILES | Cl.CC(C)NCC(=O)c1ccc(NS(=O)(=O)C)cc1 |
| IUPAC Name | N-[4-[2-(propan-2-ylamino)acetyl]phenyl]methanesulfonamide;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto