missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(4-Hydroxyphenyl)propanamide (N-Propionyl-4-aminophenol), Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
N-(4-Hydroxyphenyl)propanamide (N-Propionyl-4-aminophenol)
Specifications
Specifications
| Chemical Name or Material | 4-Propionamidophenol |
| CAS | 1693-37-4 |
| Analyte or Component Names | 4-Propionamidophenol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C9 H11 N O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Paracetamol Imp. B (EP), Acetaminophen USP Related Compound B, Acetaminophen USP RC B, N-(4-Hydroxyphenyl)-propanamide, N-Propionyl-4-aminophenol, Acetaminophen Related Compound B, Paracetamol Impurity B |
| InChI Formula | InChI=1S/C9H11NO2/c1-2-9(12)10-7-3-5-8(11)6-4-7/h3-6,11H,2H2,1H3,(H,10,12) |
| SMILES | CCC(=O)Nc1ccc(O)cc1 |
| IUPAC Name | N-(4-hydroxyphenyl)propanamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction