Learn More
Enzo Life Sciences 25MG N-Desmethyl Clozapine
Brand:  Enzo Life Sciences BML-NS106-0025
Encompass_Preferred
25MG N-Desmethyl Clozapine
Product Code. 15175428
  • € 513.04 / 25mg
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • N-Desmethyl Clozapine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.