missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N2-Methyl-5-(3,4,5-trimethoxybenzyl)pyrimidine-2,4-diamine, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
N2-Methyl-5-(3,4,5-trimethoxybenzyl)pyrimidine-2,4-diamine
Specifications
Specifications
| Chemical Name or Material | N2-Methyl-5-(3,4,5-trimethoxybenzyl)pyrimidine-2,4-diamine |
| CAS | 213745-86-9 |
| Analyte or Component Names | N2-Methyl-5-(3,4,5-trimethoxybenzyl)pyrimidine-2,4-diamine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C15 H20 N4 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Trimethoprim Imp. A (EP), N2-Methyl-5-(3,4,5-trimethoxybenzyl)pyrimidine-2,4-diamine, Trimethoprim Impurity A |
| InChI Formula | InChI=1S/C15H20N4O3/c1-17-15-18-8-10(14(16)19-15)5-9-6-11(20-2)13(22-4)12(7-9)21-3/h6-8H,5H2,1-4H3,(H3,16,17,18,19) |
| SMILES | CNc1ncc(Cc2cc(OC)c(OC)c(OC)c2)c(N)n1 |
| IUPAC Name | 2-N-methyl-5-[(3,4,5-trimethoxyphenyl)methyl]pyrimidine-2,4-diamine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction