missing translation for 'onlineSavingsMsg'
Learn More

Noroxymorphone Hydrochloride, Mikromol™

Product Code. 17983806
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 17983806

Brand: Mikromol MM0673.030025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Noroxymorphone Hydrochloride
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Noroxymorphone Hydrochloride
CAS 52446-24-9
Analyte or Component Names Noroxymorphone Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C16 H17 N O4 . Cl H
Certifications/Compliance ISO 17025
Synonym Naloxone Hydrochloride Dihydrate Imp. A (EP), Naloxone Imp. A (EP), Noroxymorphone Hydrochloride, Naloxone Hydrochloride Dihydrate Impurity A as Hydrochloride, Naltrexone Hydrochloride Impurity B as Hydrochloride, Naloxone Impurity A as Hydrochloride, Naltrexone Impurity B as Hydrochloride
InChI Formula InChI=1S/C16H17NO4.ClH/c18-9-2-1-8-7-11-16(20)4-3-10(19)14-15(16,5-6-17-11)12(8)13(9)21-14;/h1-2,11,14,17-18,20H,3-7H2;1H/t11-,14+,15+,16-;/m1./s1
SMILES Cl.Oc1ccc2C[C@H]3NCC[C@@]45[C@@H](Oc1c24)C(=O)CC[C@@]35O
IUPAC Name (4R,4aS,7aR,12bS)-4a,9-dihydroxy-1,2,3,4,5,6,7a,13-octahydro-4,12-methanobenzofuro[3,2-e]isoquinoline-7-one;hydrochloride
Molecular Weight (g/mol) 323.77
Quantity 25 mg
Formula Weight 323.0924 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.