missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nortropine, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Nortropine
Specifications
Specifications
| Chemical Name or Material | Nortropine |
| CAS | 538-09-0 |
| Analyte or Component Names | Nortropine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C7 H13 N O |
| Certifications/Compliance | ISO 17025 |
| Synonym | (3-endo)-8-Azabicyclo[3.2.1]octan-3-ol, 8-Azabicyclo[3.2.1]octan-3-ol, endo-, Nortropine (6CI,7CI), N-Demethyltropine, NSC 72850, Nortropenol, Nortropin, Tropigenin, Tropigenine, endo-8-Azabicyclo[3.2.1]octan-3-ol, 1alphaH,5alphaH-Nortropan-3alpha-ol (8CI), 3alpha-Nortropanol |
| InChI Formula | InChI=1S/C7H13NO/c9-7-3-5-1-2-6(4-7)8-5/h5-9H,1-4H2/t5-,6+,7+ |
| SMILES | O[C@H]1C[C@H]2CC[C@@H](C1)N2 |
| IUPAC Name | (1S,5R)-8-azabicyclo[3.2.1]octan-3-ol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction