missing translation for 'onlineSavingsMsg'
Learn More
Learn More
O-Acetylfluphenazine, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
O-Acetylfluphenazine
Specifications
Specifications
| Chemical Name or Material | O-Acetylfluphenazine |
| CAS | 2021-89-8 |
| Analyte or Component Names | O-Acetylfluphenazine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H28 F3 N3 O2 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | 4-[3-[2-(Trifluoromethyl)-10H-phenothiazin-10-yl]propyl]-1-piperazineethanol 1-Acetate, 1-Piperazineethanol, 4-[3-[2-(trifluoromethyl)-10H-phenothiazin-10-yl]propyl]-, acetate (ester) (9CI), 1-Piperazineethanol, 4-[3-[2-(trifluoromethyl)phenothiazin-10-yl]propyl]-, acetate (6CI,7CI,8CI), NSC 226515 |
| InChI Formula | InChI=1S/C24H28F3N3O2S/c1-18(31)32-16-15-29-13-11-28(12-14-29)9-4-10-30-20-5-2-3-6-22(20)33-23-8-7-19(17-21(23)30)24(25,26)27/h2-3,5-8,17H,4,9-16H2,1H3 |
| SMILES | CC(=O)OCCN1CCN(CCCN2c3ccccc3Sc4ccc(cc24)C(F)(F)F)CC1 |
| IUPAC Name | 2-[4-[3-[2-(trifluoromethyl)phenothiazin-10-yl]propyl]piperazin-1-yl]ethyl acetate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction