missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Risperidone trans-N-Oxide, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Risperidone trans-N-Oxide
Specifications
Specifications
| Chemical Name or Material | Risperidone trans-N-Oxide |
| Analyte or Component Names | Risperidone trans-N-Oxide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H27 F N4 O3 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Risperidone trans-N-Oxide |
| InChI Formula | InChI=1S/C23H27FN4O3/c1-15-18(23(29)27-10-3-2-4-21(27)25-15)9-13-28(30)11-7-16(8-12-28)22-19-6-5-17(24)14-20(19)31-26-22/h5-6,14,16H,2-4,7-13H2,1H3/t16-,28- |
| SMILES | CC1=C(CC[N@+]2([O-])CC[C@@H](CC2)c3noc4cc(F)ccc34)C(=O)N5CCCCC5=N1 |
| IUPAC Name | 3-[2-[4-(6-fluoro-1,2-benzoxazol-3-yl)-1-oxidopiperidin-1-ium-1-yl]ethyl]-2-methyl-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-4-one |
| Molecular Weight (g/mol) | 426.48 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction