missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(RS)-2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]acetic Acid Dihydrochloride, Mikromol
Click to view available options
Quantity:
25 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
(RS)-2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]acetic Acid Dihydrochloride
Specifications
Specifications
| Chemical Name or Material | (RS)-2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]acetic Acid Dihydrochloride |
| CAS | 1000690-91-4 |
| Analyte or Component Names | (RS)-2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]acetic Acid Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H21 Cl N2 O2 . 2 Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Cetirizine Dihydrochloride Imp. B (EP), Cetirizine Imp. B (EP), (RS)-2-[4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl]acetic Acid Dihydrochloride, Cetirizine Dihydrochloride Imp. B (EP) as Dihydrochloride, Cetirizine Dihydrochloride Impurity B as Dihydrochloride, Cetirizine Impurity B as Dihydrochloride |
| InChI Formula | InChI=1S/C19H21ClN2O2.2ClH/c20-17-8-6-16(7-9-17)19(15-4-2-1-3-5-15)22-12-10-21(11-13-22)14-18(23)24;;/h1-9,19H,10-14H2,(H,23,24);2*1H |
| SMILES | Cl.Cl.OC(=O)CN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3 |
| IUPAC Name | 2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]acetic acid;dihydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction