missing translation for 'onlineSavingsMsg'
Learn More

rac-Carbidopa EP Impurity C (Carbidopa USP Related Compound A), Mikromol™

Product Code. 30050615
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 30050615

Brand: Mikromol MM1501.090025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

rac-Carbidopa EP Impurity C (Carbidopa USP Related Compound A)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-O-Methylcarbidopa
CAS 85933-19-3
Analyte or Component Names 3-O-Methylcarbidopa
Shipping Condition Ice pack (-20°C)
Molecular Formula C11 H16 N2 O4
Certifications/Compliance ISO 9001
Synonym (RS)-2-Hydrazino-3-(4-hydroxy-3-methoxyphenyl)-2-methylpropanoic Acid ((RS)-3-O-Methylcarbidopa), alpha-Hydrazinyl-4-hydroxy-3-methoxy-alpha-methyl-benzenepropanoic acid, Benzenepropanoic acid, alpha-hydrazino-4-hydroxy-3-methoxy-alpha-methyl- (9CI), Hydrocinnamic acid, alpha-hydrazino-4-hydroxy-3-methoxy-alpha-methyl- (7CI), Carbidopa Related Compound A, Carbidopa Impurity C, Carbidopa Impurity C as Racemate
InChI Formula InChI=1S/C11H16N2O4/c1-11(13-12,10(15)16)6-7-3-4-8(14)9(5-7)17-2/h3-5,13-14H,6,12H2,1-2H3,(H,15,16)
SMILES COc1cc(CC(C)(NN)C(=O)O)ccc1O
IUPAC Name 2-hydrazinyl-3-(4-hydroxy-3-methoxyphenyl)-2-methylpropanoic acid
Molecular Weight (g/mol) 240.26
Quantity 25 mg
Formula Weight 240.111 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.