missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-2-Amino-5-methoxytetralin (S)-Mandelate, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
(S)-2-Amino-5-methoxytetralin (S)-Mandelate
Specifications
Specifications
| Chemical Name or Material | (S)-2-Amino-5-methoxytetralin (S)-Mandelate |
| CAS | 439133-67-2 |
| Analyte or Component Names | (S)-2-Amino-5-methoxytetralin (S)-Mandelate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C11 H15 N O . C8 H8 O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | (S)-2-Amino-5-methoxytetralin (S)-Mandelate, (aS)-(2S)-compd. with 1,2,3,4-Tetrahydro-5-methoxy-2-naphthalenamine a-Hydroxybenzeneacetic acid, compd. with (2S)-1,2,3,4-Tetrahydro-5-methoxy-2-naphthalenamine (aS)-a-Hydroxybenzeneacetic acid |
| InChI Formula | InChI=1S/C11H15NO.C8H8O3/c1-13-11-4-2-3-8-7-9(12)5-6-10(8)11;9-7(8(10)11)6-4-2-1-3-5-6/h2-4,9H,5-7,12H2,1H3;1-5,7,9H,(H,10,11)/t9-;7-/m00/s1 |
| SMILES | COc1cccc2C[C@@H](N)CCc12.O[C@H](C(=O)O)c3ccccc3 |
| IUPAC Name | (2S)-2-hydroxy-2-phenylacetic acid;(2S)-5-methoxy-1,2,3,4-tetrahydronaphthalen-2-amine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction