missing translation for 'onlineSavingsMsg'
Learn More

Simvastatin Methyl Ether, Mikromol™

Product Code. 30050832
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
This item is not returnable. View return policy

Product Code. 30050832

Brand: Mikromol MM0158.170025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Simvastatin Methyl Ether
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Simvastatin 4'-Methyl Ether
CAS 864357-88-0
Analyte or Component Names Simvastatin 4'-Methyl Ether
Shipping Condition Ice pack (-20°C)
Molecular Formula C26 H40 O5
Certifications/Compliance ISO 9001
Synonym Butanoic acid, 2,2-dimethyl-, (1S,3R,7S,8S,8aR)-1,2,3,7,8,8a-hexahydro-3,7-dimethyl-8-[2-[(2R,4R)-tetrahydro-4-methoxy-6-oxo-2H-pyran-2-yl]ethyl]-1-naphthalenyl ester, (1S,3R,7S,8S,8aR)-1,2,3,7,8,8a-Hexahydro-3,7-dimethyl-8-[2-[(2R,4R)-tetrahydro-4-methoxy-6-oxo-2H-pyran-2-yl]ethyl]-1-naphthalenyl 2,2-dimethylbutanoate, (1S,3R,7S,8S,8aR)-8-[2-[(2R,4R)-4-Methoxy-6-oxooxan-2-yl]ethyl]-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl 2,2-dimethylbutanoate
InChI Formula InChI=1S/C26H40O5/c1-7-26(4,5)25(28)31-22-13-16(2)12-18-9-8-17(3)21(24(18)22)11-10-19-14-20(29-6)15-23(27)30-19/h8-9,12,16-17,19-22,24H,7,10-11,13-15H2,1-6H3/t16-,17-,19+,20+,21-,22-,24-/m0/s1
SMILES CCC(C)(C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@H](CC(=O)O3)OC)[C@@H]12
IUPAC Name [(1S,3R,7S,8S,8aR)-8-[2-[(2R,4R)-4-methoxy-6-oxooxan-2-yl]ethyl]-3,7-dimethyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl] 2,2-dimethylbutanoate
Molecular Weight (g/mol) 432.59
Quantity 25 mg
Formula Weight 432.2876 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.