missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Succinylmonocholine Chloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Succinylmonocholine Chloride
Specifications
Specifications
| Chemical Name or Material | Succinylmonocholine Chloride |
| CAS | 5297-17-6 |
| Analyte or Component Names | Succinylmonocholine Chloride |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C9 H18 N O4 . Cl |
| Certifications/Compliance | ISO 9001 |
| Synonym | (3-carboxy-1-oxopropoxy)-N,N,N-trimethyl-ethanaminium chloride (1:1), Succinylmonocholine Chloride, Choline, chloride, hydrogen succinate, Succinic acid, monoester with choline chloride, Succinyl monocholine chloride |
| InChI Formula | InChI=1S/C9H17NO4.ClH/c1-10(2,3)6-7-14-9(13)5-4-8(11)12;/h4-7H2,1-3H3;1H |
| SMILES | [Cl-].C[N+](C)(C)CCOC(=O)CCC(=O)O |
| IUPAC Name | 2-(3-carboxypropanoyloxy)ethyl-trimethylazanium;chloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction