missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thiazol-5-ylmethyl [(1S,2S,4S)-4-Amino-1-benzyl-2-hydroxy-5-phenylpentyl]carbamate, Mikromol™
missing translation for 'orderingAttributeHoverText'
Quantity:
25 mg
missing translation for 'unitSize'
25mg
Description
Thiazol-5-ylmethyl [(1S,2S,4S)-4-Amino-1-benzyl-2-hydroxy-5-phenylpentyl]carbamate
Specifications
Specifications
| Chemical Name or Material | 2S,3S,5S-5-Amino-2-[N-[((5-thiozolyl)methoxy)carbonyl]amino] |
| CAS | 144164-11-4 |
| Analyte or Component Names | 2S,3S,5S-5-Amino-2-[N-[((5-thiozolyl)methoxy)carbonyl]amino] |
| Shipping Condition | Room Temperature |
| Molecular Formula | C23 H27 N3 O3 S |
| Certifications/Compliance | ISO 9001 |
| Synonym | Carbamic acid, N-[(1S,2S,4S)-4-amino-2-hydroxy-5-phenyl-1-(phenylmethyl)pentyl]-, 5-thiazolylmethyl ester, Carbamic acid, [(1S,2S,4S)-4-amino-2-hydroxy-5-phenyl-1-(phenylmethyl)pentyl]-, 5-thiazolylmethyl ester (9CI), Carbamic acid, [4-amino-2-hydroxy-5-phenyl-1-(phenylmethyl)pentyl]-, 5-thiazolylmethyl ester, [1S-(1R*,2R*,4R*)]-, 1,3-Thiazol-5-ylmethyl N-[(1S,2S,4S)-4-amino-1-benzyl-2-hydroxy-5-phenylpentyl]carbamate, Thiazol-5-ylmethyl N-[(1S,2S,4S)-4-Amino-1-benzyl-2-hydroxy-5-phenyl-pentyl]carbamate |
| InChI Formula | InChI=1S/C23H27N3O3S/c24-19(11-17-7-3-1-4-8-17)13-22(27)21(12-18-9-5-2-6-10-18)26-23(28)29-15-20-14-25-16-30-20/h1-10,14,16,19,21-22,27H,11-13,15,24H2,(H,26,28)/t19-,21-,22-/m0/s1 |
| SMILES | N[C@H](C[C@H](O)[C@H](Cc1ccccc1)NC(=O)OCc2cncs2)Cc3ccccc3 |
| IUPAC Name | 1,3-thiazol-5-ylmethyl N-[(2S,3S,5S)-5-amino-3-hydroxy-1,6-diphenylhexan-2-yl]carbamate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction