missing translation for 'onlineSavingsMsg'
Learn More
Learn More
trans-1-Benzyl-4-(4-fluorophenyl)piperidin-3-yl]methanol, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
trans-1-Benzyl-4-(4-fluorophenyl)piperidin-3-yl]methanol
Specifications
Specifications
| Chemical Name or Material | trans 1-Benzyl-4-(4-fluorophenyl)-3-piperidinemethanol |
| CAS | 188869-25-2 |
| Analyte or Component Names | trans 1-Benzyl-4-(4-fluorophenyl)-3-piperidinemethanol |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H22 F N O |
| Certifications/Compliance | ISO 9001 |
| Synonym | trans-1-Benzyl-4-(4-fluorophenyl)piperidin-3-yl]methanol, trans-1-Benzyl-4-(4-fluorophenyl)-3-hydroxymethylpiperidine, 3-Piperidinemethanol, 4-(4-fluorophenyl)-1-(phenylmethyl)-, (3R,4S)-rel-, 3-Piperidinemethanol, 4-(4-fluorophenyl)-1-(phenylmethyl)-, trans-, rel-(3R,4S)-4-(4-Fluorophenyl)-1-(phenylmethyl)-3-piperidinemethanol, trans-1-Benzyl-4-(4-fluorophenyl)-3-hydroxymethylpiperidine, trans-1-Benzyl-4-(4-fluorophenyl)piperidin-3-yl]methanol |
| InChI Formula | InChI=1S/C19H22FNO/c20-18-8-6-16(7-9-18)19-10-11-21(13-17(19)14-22)12-15-4-2-1-3-5-15/h1-9,17,19,22H,10-14H2/t17-,19-/m1/s1 |
| SMILES | OC[C@H]1CN(Cc2ccccc2)CC[C@@H]1c3ccc(F)cc3 |
| IUPAC Name | [(3R,4S)-1-benzyl-4-(4-fluorophenyl)piperidin-3-yl]methanol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction