Learn More
Wogonin, TRC
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Specifications
Specifications
| Chemical Name or Material | Wogonin |
| CAS | 632-85-9 |
| Molecular Formula | C16 H12 O5 |
| Quantity | 25 mg |
| Synonym | 4H-1-Benzopyran-4-one','5,7-dihydroxy-8-methoxy-2-phenyl-, Flavone','5,7-dihydroxy-8-methoxy- (7CI,8CI), Wogonin (6CI), 5,7-Dihydroxy-8-methoxy-2-phenyl-4H-1-benzopyran-4-one, 5,7-Dihydroxy-8-methoxyflavone |
| InChI Formula | InChI=1S/C16H12O5/c1-20-15-12(19)7-10(17)14-11(18)8-13(21-16(14)15)9-5-3-2-4-6-9/h2-8,17,19H,1H3 |
| SMILES | COc1c(O)cc(O)c2C(=O)C=C(Oc12)c3ccccc3 |
| IUPAC Name | 5,7-dihydroxy-8-methoxy-2-phenylchromen-4-one |
| Molecular Weight (g/mol) | 284.26 |
| Formula Weight | 284.0685 |