missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(Z)-2-Phenylbut-2-enyl 3,4,5-Trimethoxybenzoate, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
(Z)-2-Phenylbut-2-enyl 3,4,5-Trimethoxybenzoate
Specifications
Specifications
| Chemical Name or Material | (Z)-2-Phenylbut-2-enyl 3,4,5-Trimethoxybenzoate |
| Analyte or Component Names | (Z)-2-Phenylbut-2-enyl 3,4,5-Trimethoxybenzoate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H22 O5 |
| Certifications/Compliance | ISO 9001 |
| Synonym | (Z)-2-Phenylbut-2-enyl 3,4,5-Trimethoxybenzoate |
| InChI Formula | InChI=1S/C20H22O5/c1-5-14(15-9-7-6-8-10-15)13-25-20(21)16-11-17(22-2)19(24-4)18(12-16)23-3/h5-12H,13H2,1-4H3/b14-5+ |
| SMILES | COc1cc(cc(OC)c1OC)C(=O)OC\C(=C/C)\c2ccccc2 |
| IUPAC Name | [(Z)-2-phenylbut-2-enyl] 3,4,5-trimethoxybenzoate |
| Molecular Weight (g/mol) | 342.39 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction