Learn More
Merck Accessories for Glass Microanalysis Filter Holders
Accessories for filter holders for sample preparation.
Brand: Merck XX1012532
440.63 EUR valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Additional Details : Weight : 0.84060kg
Description
Accessories for filter holders for sample preparation.
Specifications
Not Autoclavable | |
25 mm glass microanalysis vacuum filter | |
Non-sterile | |
25 mm |
Base, stopper and screen | |
Borosilicate Glass | |
Base, Stopper and Screen |