missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Caspase-10 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB834-SP
This item is not returnable.
View return policy
Description
Caspase-10/FLICE2 Monoclonal specifically detects Caspase-10/FLICE2 in Human samples. It is validated for Western Blot.
Specifications
| Caspase-10/FLICE2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| AAC50644 | |
| CASP10 | |
| E. coli-derived recombinant human Caspase-10/a/B (Mch 4) Val220-Ile479 Accession # AAC50644 | |
| 25 μg | |
| Primary | |
| Detects human Caspase-10 in Western blots. | |
| Human | |
| Purified |
| Western Blot | |
| 63131 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Apoptotic protease Mch-4, CASP10, CASP-10, caspase 10, apoptosis-related cysteine peptidase, caspase 10, apoptosis-related cysteine protease, caspase-10, Caspase10, EC 3.4.22.63, FADD-like ICE2, FAS-associated death domain protein interleukin-1B-converting enzyme 2, FLICE2, ICE-like apoptotic protease 4, interleukin-1B-converting enzyme 2, Mch4, MCH4ALPS2 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 843 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction