missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Coagulation Factor II/Thrombin Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF1148-SP
This item is not returnable.
View return policy
Description
Coagulation Factor II/Thrombin Polyclonal specifically detects Coagulation Factor II/Thrombin in Human samples. It is validated for Western Blot, Simple Western, Flow Cytometry, CyTOF-ready.
Specifications
| Coagulation Factor II/Thrombin | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| F2 | |
| Mouse myeloma cell line NS0-derived recombinant human Coagulation Factor II/Thrombin | |
| 25 μg | |
| Primary | |
| Detects human Coagulation Factor II/Thrombin in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) PROC is observed, and less than 1% cross-reactivity with rhFactor X, recombinant mouse (rm) Factor X and rmPROC is observed. | |
| Human | |
| IgG |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Simple Western 10 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Coagulation Factor II, coagulation factor II (thrombin) receptor-like 2, Coagulation factor II receptor-like 2, Coagulation factor II receptor-like 2 (protease-actovated receptor 3), F2, PAR-3, PAR3proteinase-activated receptor 3, protease-activated receptor 3, proteinase-activated receptor-3, PT, serine protease, Thrombin, Thrombin receptor-like 2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2147 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction