missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GGT5 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7209-SP
This item is not returnable.
View return policy
Description
GGT5 Polyclonal specifically detects GGT5 in Human samples. It is validated for Western Blot.
Specifications
| GGT5 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp566O011, EC 2.3.2.2, gamma-glutamyl cleaving enzyme, Gamma-glutamyl transpeptidase-related enzyme, gamma-glutamyl transpeptidase-related protein, gamma-glutamyltransferase 5, Gamma-glutamyltransferase-like activity 1FLJ92733, Gamma-glutamyltranspeptidase 5, GGT 5, GGTLA1, GGT-rel | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 2687 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P36269 | |
| GGT5 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human GGT5 Ser30-Tyr586 Accession # P36269 | |
| 25 μg | |
| Primary | |
| Detects human GGT5 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction