missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse ICAM-2/CD102 Antibody, R&D Systems™

Description
ICAM-2/CD102 Polyclonal specifically detects ICAM-2/CD102 in Mouse samples. It is validated for Western Blot, Immunohistochemistry, Adhesion Blockade.
Specifications
Specifications
| Antigen | ICAM-2/CD102 |
| Applications | Western Blot, Immunohistochemistry, Adhesion Blockade |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.25 ug/mL, Immunohistochemistry 5-15 ug/mL, Adhesion Blockade |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | P35330 |
| Gene Alias | CD102, CD102 antigen, ICAM2, ICAM-2, intercellular adhesion molecule 2 |
| Gene Symbols | ICAM2 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?