missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LPPR2 Antibody [Unconjugated], Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-43685-25ul
This item is not returnable.
View return policy
Description
LPPR2 Polyclonal antibody specifically detects LPPR2 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
Specifications
| LPPR2 | |
| Polyclonal | |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| DKFZp761E1121, EC 3.1.3.4, FLJ13055, lipid phosphate phosphatase-related protein type 2, plasticity related gene 4, Plasticity-related gene 4 protein, PRG4, PRG-4 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: YTALGCLPPSPDRPGPDRFVTDQGACAGSPSLVAAARRAFPCKDAA | |
| 25 μL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 64748 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction