missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2A Peptide Antibody (3H4), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59627JF549
This item is not returnable.
View return policy
Description
2A Peptide Monoclonal antibody specifically detects 2A Peptide in Nonspecies specific samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Specifications
| 2A Peptide | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 2A cleavage peptide, 2A Peptide, 2A peptide tag, 2A sequence, 2A-like peptide, antibody for 2A-containing recombinant proteins, FMDV 2A-peptide, MDV 2A-peptide, P2A peptide, P2A tagged proteins, peptide 2A sequences, self-cleaving 2A peptide, T2A peptide, T2A tagged proteins, Thosea asigna virus 2A | |
| This 2A Peptide Antibody (3H4) was developed against KLH-coupled synthetic peptide corresponding to 2A peptide of Thosea asigna virus (CGDVEENPG) | |
| 0.1 mL | |
| Epitope Tags, Virology Bacteria and Parasites | |
| Non-species specific | |
| Purified |
| Western Blot, Immunofluorescence, Immunoprecipitation | |
| 3H4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction