missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2B4/CD244/SLAMF4 Antibody (146506), Allophycocyanin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB10392APC
This item is not returnable.
View return policy
Description
2B4/CD244/SLAMF4 Monoclonal specifically detects 2B4/CD244/SLAMF4 in Human samples. It is validated for Flow Cytometry.
Specifications
| 2B4/CD244/SLAMF4 | |
| Monoclonal | |
| APC | |
| PBS with 0.05% Sodium Azide | |
| CD244 | |
| Mouse myeloma cell line NS0-derived partial recombinant human 2B4/CD244/SLAMF4 protein (amino acids 22-221) | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| 146506 | |
| Flow Cytometry | |
| CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4NK cell type I receptor protein 2B4,2B4CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4NK cell activation inducing ligand NAIL | |
| Mouse | |
| Protein A or G purified | |
| Myeloid derived Suppressor Cell, Phospho Specific | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction