missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2B4/CD244/SLAMF4 Rabbit anti-Human, Clone: 25, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 367.00 - € 552.00
Specifications
| Antigen | 2B4/CD244/SLAMF4 |
|---|---|
| Clone | 25 |
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1-4 ul/mg of lysate, Sandwich ELISA Capture 1:250-1:2000 |
| Applications | Western Blot, Immunoprecipitation, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18227086
|
Novus Biologicals
NBP2-89232-50UL |
50 μg |
€ 367.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18277047
|
Novus Biologicals
NBP2-89232-100UL |
100 μg |
€ 552.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2B4/CD244/SLAMF4 Monoclonal specifically detects 2B4/CD244/SLAMF4 in Human samples. It is validated for Western Blot, Immunoprecipitation, Sandwich ELISA Capture.Specifications
| 2B4/CD244/SLAMF4 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1-4 ul/mg of lysate, Sandwich ELISA Capture 1:250-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD244 antigen, CD244 molecule, natural killer cell receptor 2B4, h2B4, NAILNK cell activation-inducing ligand, natural killer cell receptor 2B4, NKR2B4NK cell type I receptor protein 2B4,2B4CD244 natural killer cell receptor 2B4, Nmrk, SLAMF4NK cell activation inducing ligand NAIL | |
| Recombinant Human 2B4/CD244/SLAMF4 protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 25 | |
| Western Blot, Immunoprecipitation, ELISA | |
| Unconjugated | |
| Rabbit | |
| Myeloid derived Suppressor Cell, Phospho Specific | |
| 0.2 um filtered solution in PBS | |
| 51744 | |
| IgG | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title