missing translation for 'onlineSavingsMsg'
Learn More

(2E)-4-Methoxy-2-butenoic Acid, TRC

Product Code. 30468873
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
Unit Size:
1g
2.5g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30468873 2.5 g 2.5g
30479448 1 g 1g
30481589 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30468873 Supplier Toronto Research Chemicals Supplier No. TRCM2623702.5G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63968-74-1
Molecular Formula C5 H8 O3
Molecular Weight (g/mol) 116.12
Synonym (E)-4-Methoxy-2-butenoic Acid
IUPAC Name (E)-4-methoxybut-2-enoic acid
SMILES COC\C=C\C(=O)O

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C5H8O3/c1-8-4-2-3-5(6)7/h2-3H,4H2,1H3,(H,6,7)/b3-2+
Formula Weight 116.0473
Chemical Name or Material (2E)-4-Methoxy-2-butenoic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.