missing translation for 'onlineSavingsMsg'
Learn More

(2E)-N,6,6-Trimethyl-N-[(4-methylnaphthalen-1-yl)methyl]hept-2-en-4-yn-1-amine Hydrochloride (4-Methylterbinafine Hydrochloride), Mikromol™

Product Code. 30050301
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30050301 25 mg 25mg
30052729 100 mg 100mg
30460982 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30050301 Supplier Mikromol Supplier No. MM0696.050025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2E)-N,6,6-Trimethyl-N-[(4-methylnaphthalen-1-yl)methyl]hept-2-en-4-yn-1-amine Hydrochloride (4-Methylterbinafine Hydrochloride)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methylterbinafine Hydrochloride
CAS 877265-33-3
Analyte or Component Names 4-Methylterbinafine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C22 H27 N . Cl H
Certifications/Compliance ISO 9001
Synonym Terbinafine Hydrochloride Imp. D (EP), Terbinafine Imp. D (EP), Terbinafine USP Related Compound D, Terbinafine USP RC D, (2E)-N,6,6-Trimethyl-N-[(4-methylnaphthalen-1-yl)methyl]-hept-2-en-4-yn-1-amine Hydrochloride, Terbinafine Hydrochloride Imp. D (EP) as Hydrochloride, 4-Methylterbinafine Hydrochloride, Terbinafine Related Compound D as Hydrochloride, Terbinafine Hydrochloride Impurity D as Hydrochloride, Terbinafine Impurity D as Hydrochloride
InChI Formula InChI=1S/C22H27N.ClH/c1-18-13-14-19(21-12-8-7-11-20(18)21)17-23(5)16-10-6-9-15-22(2,3)4;/h6-8,10-14H,16-17H2,1-5H3;1H/b10-6+;
SMILES Cl.CN(C\C=C\C#CC(C)(C)C)Cc1ccc(C)c2ccccc12
IUPAC Name (E)-N,6,6-trimethyl-N-[(4-methylnaphthalen-1-yl)methyl]hept-2-en-4-yn-1-amine;hydrochloride
Molecular Weight (g/mol) 341.917
Quantity 25 mg
Formula Weight 341.191 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.