missing translation for 'onlineSavingsMsg'
Learn More

(2E)-N,6,6-Trimethyl-N-[(4-methylnaphthalen-1-yl)methyl]hept-2-en-4-yn-1-amine Hydrochloride (4-Methylterbinafine Hydrochloride), Mikromol™

Product Code. 30460982
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30460982 4 x 25 mg 100mg
30050301 25 mg 25mg
30052729 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30460982 Supplier Mikromol Supplier No. MM0696.0500254AMP
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2E)-N,6,6-Trimethyl-N-[(4-methylnaphthalen-1-yl)methyl]hept-2-en-4-yn-1-amine Hydrochloride (4-Methylterbinafine Hydrochloride), Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-Methylterbinafine Hydrochloride
CAS 877265-33-3
Analyte or Component Names 4-Methylterbinafine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C22 H27 N . Cl H
Certifications/Compliance ISO 9001
Synonym Terbinafine Hydrochloride Imp. D (EP), Terbinafine Imp. D (EP), Terbinafine USP Related Compound D, Terbinafine USP RC D, (2E)-N,6,6-Trimethyl-N-[(4-methylnaphthalen-1-yl)methyl]-hept-2-en-4-yn-1-amine Hydrochloride, Terbinafine Hydrochloride Imp. D (EP) as Hydrochloride, 4-Methylterbinafine Hydrochloride, Terbinafine Related Compound D as Hydrochloride, Terbinafine Hydrochloride Impurity D as Hydrochloride, Terbinafine Impurity D as Hydrochloride
InChI Formula InChI=1S/C22H27N.ClH/c1-18-13-14-19(21-12-8-7-11-20(18)21)17-23(5)16-10-6-9-15-22(2,3)4;/h6-8,10-14H,16-17H2,1-5H3;1H/b10-6+;
Solution Type Neat
SMILES Cl.CN(C\C=C\C#CC(C)(C)C)Cc1ccc(C)c2ccccc12
IUPAC Name (E)-N,6,6-trimethyl-N-[(4-methylnaphthalen-1-yl)methyl]hept-2-en-4-yn-1-amine;hydrochloride
Molecular Weight (g/mol) 341.917
Quantity 4 x 25 mg
Formula Weight 341.191 g/mol
Afficher plus Afficher moins
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.