missing translation for 'onlineSavingsMsg'
Learn More

(2E)-N,6,6-Trimethyl-N-(naphthalen-2-ylmethyl)hept-2-en-4-yn-1-amine Hydrochloride (trans-Isoterbinafine Hydrochloride), Mikromol™

Product Code. 17903576
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17903576 25 mg 25mg
17902776 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17903576 Supplier Mikromol Supplier No. MM0696.040025

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2E)-N,6,6-Trimethyl-N-(naphthalen-2-ylmethyl)hept-2-en-4-yn-1-amine Hydrochloride (trans-Isoterbinafine Hydrochloride)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material trans-Isoterbinafine Hydrochloride
CAS 877265-30-0
Analyte or Component Names trans-Isoterbinafine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C21 H25 N . Cl H
Certifications/Compliance ISO 17025
Synonym Terbinafine Hydrochloride Imp. C (EP), Terbinafine Imp. C (EP), Terbinafine USP Related Compound C, Terbinafine USP RC C, (2E)-N,6,6-Trimethyl-N-(naphthalen-2-ylmethyl)hept-2-en-4-yn-1-amine Hydrochloride, Terbinafine Hydrochloride Imp. C (EP) as Hydrochloride, trans-Isoterbinafine Hydrochloride, Terbinafine Related Compound C as Hydrochloride, Terbinafine Hydrochloride Impurity C as Hydrochloride, Terbinafine Impurity C as Hydrochloride
InChI Formula InChI=1S/C21H25N.ClH/c1-21(2,3)14-8-5-9-15-22(4)17-18-12-13-19-10-6-7-11-20(19)16-18;/h5-7,9-13,16H,15,17H2,1-4H3;1H/b9-5+;
SMILES Cl.CN(C\C=C\C#CC(C)(C)C)Cc1ccc2ccccc2c1
IUPAC Name (E)-N,6,6-trimethyl-N-(naphthalen-2-ylmethyl)hept-2-en-4-yn-1-amine;hydrochloride
Molecular Weight (g/mol) 327.89
Quantity 25 mg
Formula Weight 327.1754 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.