Learn More
(2RS)-2-Amino-4-phenylbutanoic Acid, Mikromol™
Product Code. 17989104
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. 17989104
Brand: Mikromol MM0598.01
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
(2RS)-2-Amino-4-phenylbutanoic Acid
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material (2RS)-2-Amino-4-phenylbutanoic Acid
CAS 1012-05-1
Analyte or Component Names (2RS)-2-Amino-4-phenylbutanoic Acid
Shipping Condition Room Temperature
Molecular Formula C10 H13 N O2
Certifications/Compliance ISO 17025
Synonym Lisinopril Dihydrate Imp. A (EP), Lisinopril Imp. A (EP), (2RS)-2-Amino-4-phenylbutanoic Acid, Lisinopril Dihydrate Impurity A, Lisinopril Impurity A
InChI Formula InChI=1S/C10H13NO2/c11-9(10(12)13)7-6-8-4-2-1-3-5-8/h1-5,9H,6-7,11H2,(H,12,13)
SMILES NC(CCc1ccccc1)C(=O)O
IUPAC Name 2-amino-4-phenylbutanoic acid
Molecular Weight (g/mol) 179.22
Quantity 100 mg
Formula Weight 179.0946 g/mol