missing translation for 'onlineSavingsMsg'
Learn More

(2RS)-2-Methyl-3-sulfanyl-propanoic Acid, Mikromol™

Product Code. 30051708
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051708 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30051708 Supplier Mikromol Supplier No. MM0067.02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(2RS)-2-Methyl-3-sulfanyl-propanoic Acid
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2RS)-2-Methyl-3-sulfanyl-propanoic Acid
CAS 26473-47-2
Analyte or Component Names (2RS)-2-Methyl-3-sulfanyl-propanoic Acid
Shipping Condition Ice pack (-20°C)
Molecular Formula C4 H8 O2 S
Certifications/Compliance ISO 9001
Synonym (2RS)-2-Methyl-3-sulfanyl-propanoic Acid, Captopril Imp. C (EP), Propionic acid, 3-mercapto-2-methyl- (8CI), 3-Mercapto-2-methylpropanoic acid, 3-Mercapto-2-methylpropionic acid, 3-Mercaptoisobutyric acid, Isobutyric acid, beta-mercapto- (4CI), Captopril Impurity C
InChI Formula InChI=1S/C4H8O2S/c1-3(2-7)4(5)6/h3,7H,2H2,1H3,(H,5,6)
SMILES CC(CS)C(=O)O
IUPAC Name 2-methyl-3-sulfanylpropanoic acid
Molecular Weight (g/mol) 120.17
Quantity 100 mg
Formula Weight 120.0245 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.